4-Bromo-2-fluoro-6-methoxybenzamide ≥95%

Supplier: AOBChem USA

Warning

59056-1G 59056-500MG 59056-5G 59056-25G
77472-084EA 310.03 USD
77472-084 77484-710 77495-246 77504-646
4-Bromo-2-fluoro-6-methoxybenzamide ≥95%
4-Bromo-2-fluoro-6-methoxybenzamide

Precaución: For research use only

Formula: C₈H₇BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD30468975
CAS Number: 2092532-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR