1-(2,4-Bis(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

58999-1G 58999-500MG 58999-5G 58999-25G
77472-080EA 253.38 USD
77472-080 77484-706 77495-242 77504-644
1-(2,4-Bis(trifluoromethyl)phenyl)ethanol ≥95%
1-(2,4-Bis(trifluoromethyl)phenyl)ethanol

Precaución: For research use only

Formula: C₁₀H₈F₆O
MW: 258.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25977622
CAS Number: 1265177-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR