(2-Chloro-6-fluoro-3-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58997-1G 58997-500MG 58997-5G
77472-076EA 832.24 USD
77472-076 77484-702 77495-238
(2-Chloro-6-fluoro-3-iodophenyl)methanol ≥97%
(2-Chloro-6-fluoro-3-iodophenyl)methanol

Precaución: For research use only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758945
CAS Number: 2383989-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR