2-(Benzyloxy)-1-bromo-3,4-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

58873-1G 58873-500MG 58873-5G
77472-072EA 1085.62 USD
77472-072 77484-698 77495-234
2-(Benzyloxy)-1-bromo-3,4-dimethoxybenzene ≥95%
2-(Benzyloxy)-1-bromo-3,4-dimethoxybenzene

Precaución: For research use only

Formula: C₁₅H₁₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD34182893
CAS Number: 1335227-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR