2-(5-Bromo-2-ethoxy-3-methoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

58603-1G 58603-500MG 58603-5G
77472-068EA 815.76 USD
77472-068 77484-694 77495-230
2-(5-Bromo-2-ethoxy-3-methoxyphenyl)-1,3-dioxolane ≥95%
2-(5-Bromo-2-ethoxy-3-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₂H₁₅BrO₄
MW: 303.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758724
CAS Number: 2755717-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR