4-(Methoxymethoxy)-2,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58450-1G 58450-500MG 58450-5G 58450-25G
77472-064EA 415.09 USD
77472-064 77484-690 77495-224 77504-638
4-(Methoxymethoxy)-2,5-dimethylbenzaldehyde ≥97%
4-(Methoxymethoxy)-2,5-dimethylbenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27997616
CAS Number: 170930-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR