1-Bromo-4-(methoxymethoxy)-2,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

58446-1G 58446-5G 58446-25G
77472-060EA 64.89 USD
77472-060 77495-220 77504-634
1-Bromo-4-(methoxymethoxy)-2,5-dimethylbenzene ≥95%
1-Bromo-4-(methoxymethoxy)-2,5-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD27997615
CAS Number: 1823953-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR