(5-Fluoro-4-methoxy-2-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

58437-1G 58437-500MG
77472-056EA 3422.69 USD
77472-056 77484-684
(5-Fluoro-4-methoxy-2-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(5-Fluoro-4-methoxy-2-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₈F₄OS
MW: 240.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758674
CAS Number: 2708458-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR