2-Methyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

58315-1G 58315-500MG 58315-5G 58315-25G
77472-052EA 379.04 USD
77472-052 77484-680 77495-216 77504-630
2-Methyl-[1,1'-biphenyl]-3-amine ≥97%
2-Methyl-[1,1'-biphenyl]-3-amine

Precaución: For research use only

Formula: C₁₃H₁₃N
MW: 183.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14585563
CAS Number: 172975-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR