4-Ethoxy-3-fluoro-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58174-1G 58174-500MG 58174-5G 58174-25G
77472-048EA 399.64 USD
77472-048 77484-676 77495-212 77504-626
4-Ethoxy-3-fluoro-2-(methylthio)benzaldehyde ≥97%
4-Ethoxy-3-fluoro-2-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁FO₂S
MW: 214.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758501
CAS Number: 2901066-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR