3-Bromo-6-chloro-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

58154-1G 58154-500MG 58154-5G 58154-25G
77472-044EA 166.86 USD
77472-044 77484-672 77495-208 77504-622
3-Bromo-6-chloro-2-methylphenol ≥97%
3-Bromo-6-chloro-2-methylphenol

Precaución: For research use only

Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD34758488
CAS Number: 2092417-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR