2,3-Dichloro-6-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58010-1G 58010-500MG 58010-5G
77472-040EA 829.15 USD
77472-040 77484-668 77495-204
2,3-Dichloro-6-iodobenzaldehyde ≥97%
2,3-Dichloro-6-iodobenzaldehyde

Precaución: For research use only

Formula: C₇H₃Cl₂IO
MW: 300.909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758422
CAS Number: 2092867-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR