3-Bromo-2-chloro-5-ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57877-1G 57877-500MG 57877-5G
77472-036EA 515 USD
77472-036 77484-664 77495-200
3-Bromo-2-chloro-5-ethylbenzaldehyde ≥97%
3-Bromo-2-chloro-5-ethylbenzaldehyde

Precaución: For research use only

Formula: C₉H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD28505590
CAS Number: 1935490-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR