Ethyl 3-hydroxy-3-(thiophen-2-yl)propanoate ≥95%

Supplier: AOBChem USA

Warning

57800-1G 57800-500MG 57800-5G
77472-032EA 654.05 USD
77472-032 77484-662 77495-196
Ethyl 3-hydroxy-3-(thiophen-2-yl)propanoate ≥95%
Ethyl 3-hydroxy-3-(thiophen-2-yl)propanoate

Precaución: For research use only

Formula: C₉H₁₂O₃S
MW: 200.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16749057
CAS Number: 40029-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR