Methyl 5-bromo-3-(tert-butyl)-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

57747-1G 57747-500MG 57747-5G
77472-028EA 653.02 USD
77472-028 77484-658 77495-192
Methyl 5-bromo-3-(tert-butyl)-2-methoxybenzoate ≥95%
Methyl 5-bromo-3-(tert-butyl)-2-methoxybenzoate

Precaución: For research use only

Formula: C₁₃H₁₇BrO₃
MW: 301.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758287
CAS Number: 2121251-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR