3'-Bromo-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57573-1G 57573-500MG 57573-5G
77472-024EA 515 USD
77472-024 77484-654 77495-188
3'-Bromo-2-methyl-1,1'-biphenyl ≥97%
3'-Bromo-2-methyl-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₃H₁₁Br
Storage Temperature: Ambient
MDL Number: MFCD26632021
CAS Number: 864149-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR