Methyl 6-ethoxy-3-fluoro-2-(Methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

57521-1G 57521-500MG 57521-5G 57521-25G
77472-020EA 298.7 USD
77472-020 77484-650 77495-184 77504-612
Methyl 6-ethoxy-3-fluoro-2-(Methylthio)benzoate ≥97%
Methyl 6-ethoxy-3-fluoro-2-(Methylthio)benzoate

Precaución: For research use only

Formula: C₁₁H₁₃FO₃S
MW: 244.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758218
CAS Number: 2755723-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR