1-Bromo-2,3-dimethoxy-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

57501-1G 57501-500MG 57501-5G
77472-016EA 676.71 USD
77472-016 77484-646 77495-180
1-Bromo-2,3-dimethoxy-5-methylbenzene ≥95%
1-Bromo-2,3-dimethoxy-5-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22414740
CAS Number: 68278-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR