4-Bromo-2-fluoro-3-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

57488-1G 57488-500MG 57488-5G
77472-012EA 482.04 USD
77472-012 77484-642 77495-176
4-Bromo-2-fluoro-3-methoxy-5-methylbenzoic acid ≥97%
4-Bromo-2-fluoro-3-methoxy-5-methylbenzoic acid

Precaución: For research use only

Formula: C₉H₈BrFO₃
MDL Number: MFCD34758204
CAS Number: 1891540-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR