1-Bromo-4-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥97%

Supplier: AOBChem USA

57021-5G 57021-25G
77495-156EA 912.58 USD
77495-156 77504-596
1-Bromo-4-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥97%
1-Bromo-4-fluoro-2-(methoxymethoxy)-3-methylbenzene

Precaución: For research use only

Formula: C₉H₁₀BrFO₂
MDL Number: MFCD34758022
CAS Number: 2386648-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR