2,2-Dichloro-1-(4-ethoxy-3-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

56541-5G 56541-25G
77495-118EA 1484.23 USD
77495-118 77504-566
2,2-Dichloro-1-(4-ethoxy-3-fluorophenyl)ethanol ≥95%
2,2-Dichloro-1-(4-ethoxy-3-fluorophenyl)ethanol

Precaución: For research use only

Formula: C₁₀H₁₁Cl₂FO₂
MW: 253.098 g/mol
MDL Number: MFCD34813915
CAS Number: 2921719-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR