2-Bromo-4-fluoro-3-iodo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

56124-5G 56124-25G
77495-094EA 227.63 USD
77495-094 77504-552
2-Bromo-4-fluoro-3-iodo-1-methoxybenzene ≥97%
2-Bromo-4-fluoro-3-iodo-1-methoxybenzene

Precaución: For research use only

Formula: C₇H₅BrFIO
MDL Number: MFCD34530486
CAS Number: 2384800-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR