1-Bromo-5-fluoro-4-iodo-2-methoxybenzene ≥97%

Supplier: AOBChem USA

56044-5G 56044-25G
77495-086EA 1484.23 USD
77495-086 77504-546
1-Bromo-5-fluoro-4-iodo-2-methoxybenzene ≥97%
1-Bromo-5-fluoro-4-iodo-2-methoxybenzene

Precaución: For research use only

Formula: C₇H₅BrFIO
MDL Number: MFCD27975440
CAS Number: 677777-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR