(9-Isopropyl-9H-carbazol-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

55886-5G 55886-25G
77495-078EA 1484.23 USD
77495-078 77504-538
(9-Isopropyl-9H-carbazol-3-yl)boronic acid ≥97%
(9-Isopropyl-9H-carbazol-3-yl)boronic acid

Precaución: For research use only

Formula: C₁₅H₁₆BNO₂
MDL Number: MFCD34757629
CAS Number: 2308494-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR