3-(Methylthio)benzo[b]thiophene-2-carbaldehyde ≥95%

Supplier: AOBChem USA

55717-5G 55717-25G
77495-070EA 1484.23 USD
77495-070 77504-530
3-(Methylthio)benzo[b]thiophene-2-carbaldehyde ≥95%

Precaución: For research use only

Formula: C₁₀H₈OS₂
MDL Number: MFCD00465000
CAS Number: 50451-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR