1-Fluoro-4-methoxy-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

55588-1G 55588-500MG 55588-5G 55588-25G
77471-900EA 120.51 USD
77471-900 77484-536 77495-058 77504-520
1-Fluoro-4-methoxy-2-(methoxymethoxy)benzene ≥95%
1-Fluoro-4-methoxy-2-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₉H₁₁FO₃
MW: 186.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564310
CAS Number: 2120398-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR