3-Chloro-5-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

55537-1G 55537-500MG 55537-5G 55537-25G
77471-896EA 415.09 USD
77471-896 77484-532 77495-054 77504-516
3-Chloro-5-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3-Chloro-5-fluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: For research use only

Formula: C₁₃H₈ClFO
MW: 234.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757536
CAS Number: 2901096-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR