1-([1,1'-Biphenyl]-4-yl)-2,2-dichloroethanol ≥95%

Supplier: AOBChem USA

Warning

55255-1G 55255-500MG 55255-5G 55255-25G
77471-888EA 82.4 USD
77471-888 77484-524 77495-044 77504-506
1-([1,1'-Biphenyl]-4-yl)-2,2-dichloroethanol ≥95%
1-([1,1'-Biphenyl]-4-yl)-2,2-dichloroethanol

Precaución: For research use only

Formula: C₁₄H₁₂Cl₂O
MW: 267.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02689980
CAS Number: 59612-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR