2,2-Dichloro-1-(o-tolyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

54976-1G 54976-500MG 54976-5G 54976-25G
77471-880EA 111.24 USD
77471-880 77484-516 77495-036 77504-500
2,2-Dichloro-1-(o-tolyl)ethanol ≥95%
2,2-Dichloro-1-(o-tolyl)ethanol

Precaución: For research use only

Formula: C₉H₁₀Cl₂O
MW: 205.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530332
CAS Number: 18067-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR