Methyl 2-fluoro-4-formyl-3-Methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

54884-1G 54884-500MG 54884-5G 54884-25G
77471-876EA 221.45 USD
77471-876 77484-512 77495-032 77504-496
Methyl 2-fluoro-4-formyl-3-Methylbenzoate ≥97%
Methyl 2-fluoro-4-formyl-3-Methylbenzoate

Precaución: For research use only

Formula: C₁₀H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD34757407
CAS Number: 2384987-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR