1-Bromo-2,5-dichloro-4-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

54761-1G 54761-500MG 54761-5G 54761-25G
77471-872EA 253.38 USD
77471-872 77484-508 77495-028 77504-492
1-Bromo-2,5-dichloro-4-(methoxymethoxy)benzene ≥95%
1-Bromo-2,5-dichloro-4-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇BrCl₂O₂
MW: 285.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564275
CAS Number: 1301147-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR