4-Bromo-6-methyl-2-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

54722-1G 54722-500MG 54722-5G 54722-25G
77471-868EA 180.25 USD
77471-868 77484-504 77495-024 77504-488
4-Bromo-6-methyl-2-methoxyphenol ≥95%
4-Bromo-6-methyl-2-methoxyphenol

Precaución: For research use only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD09263254
CAS Number: 86232-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR