5-Bromo-2-isopropoxy-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54678-1G 54678-500MG 54678-5G 54678-25G
77471-864EA 393.46 USD
77471-864 77484-500 77495-020 77504-486
5-Bromo-2-isopropoxy-3-methoxybenzaldehyde ≥97%
5-Bromo-2-isopropoxy-3-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₃BrO₃
MW: 273.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28641690
CAS Number: 101417-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR