(2-Formylbenzo[b]thiophen-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

54336-500MG 54336-5G 54336-25G
77484-476EA 263.68 USD
77484-476 77494-996 77504-466
(2-Formylbenzo[b]thiophen-3-yl)boronic acid ≥97%
(2-Formylbenzo[b]thiophen-3-yl)boronic acid

Precaución: For research use only

Formula: C₉H₇BO₃S
Storage Temperature: Ambient
MDL Number: MFCD18383161
CAS Number: 177735-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR