2-Chloro-4-fluoro-6-methylphenol ≥95%

Supplier: AOBChem USA

Warning

54215-500MG 54215-5G 54215-25G
77484-472EA 141.11 USD
77484-472 77494-992 77504-462
2-Chloro-4-fluoro-6-methylphenol ≥95%
2-Chloro-4-fluoro-6-methylphenol

Precaución: For research use only

Formula: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD22415117
CAS Number: 1804404-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR