3-(Methoxymethoxy)-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

53982-500MG 53982-5G 53982-25G
77484-468EA 141.11 USD
77484-468 77494-984 77504-454
3-(Methoxymethoxy)-5-methylbenzoic acid ≥95%
3-(Methoxymethoxy)-5-methylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757208
CAS Number: 2807445-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR