5-(tert-Butyl)-2-chlorophenol ≥95%

Supplier: AOBChem USA

Warning

53778-500MG 53778-5G
77484-460EA 349.17 USD
77484-460 77494-972
5-(tert-Butyl)-2-chlorophenol ≥95%
5-(tert-Butyl)-2-chlorophenol

Precaución: For research use only

Formula: C₁₀H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD16996870
CAS Number: 20942-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR