(4-Butoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

53697-500MG 53697-5G
77484-456EA 463.5 USD
77484-456 77494-966
(4-Butoxyphenyl)(methyl)sulfane ≥95%
(4-Butoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD22373591
CAS Number: 1443310-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR