3-(tert-Butyl)-5-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

53669-500MG 53669-5G 53669-25G
77484-452EA 296.64 USD
77484-452 77494-962 77504-442
3-(tert-Butyl)-5-(methylthio)benzaldehyde ≥95%
3-(tert-Butyl)-5-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₁₂H₁₆OS
MW: 208.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29122096
CAS Number: 1289111-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR