1-(3-Chloro-5-(trifluoromethyl)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

53608-500MG 53608-5G 53608-25G
77484-448EA 161.71 USD
77484-448 77494-958 77504-438
1-(3-Chloro-5-(trifluoromethyl)phenyl)ethanol ≥97%
1-(3-Chloro-5-(trifluoromethyl)phenyl)ethanol

Precaución: For research use only

Formula: C₉H₈ClF₃O
MW: 224.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22572643
CAS Number: 1252657-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR