2-(2-Bromo-4-fluoro-5-methylphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

53550-500MG 53550-5G 53550-25G
77484-440EA 127.72 USD
77484-440 77494-950 77504-430
2-(2-Bromo-4-fluoro-5-methylphenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-4-fluoro-5-methylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757065
CAS Number: 2807454-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR