5-Bromo-2-fluoro-6-methylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

53526-500MG 53526-5G 53526-25G
77484-436EA 128.75 USD
77484-436 77494-946 77504-426
5-Bromo-2-fluoro-6-methylnicotinic acid ≥97%
5-Bromo-2-fluoro-6-methylnicotinic acid

Precaución: For research use only

Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD34757058
CAS Number: 2764728-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR