2,5-Dimethyl-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

53484-500MG 53484-5G 53484-25G
77484-432EA 296.64 USD
77484-432 77494-942 77504-422
2,5-Dimethyl-3-(methylthio)benzaldehyde ≥97%
2,5-Dimethyl-3-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₂OS
MW: 180.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757045
CAS Number: 2901096-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR