6-Chloro-2-fluoro-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

53415-500MG 53415-5G 53415-25G
77484-428EA 60.77 USD
77484-428 77494-936 77504-418
6-Chloro-2-fluoro-3-(methylthio)pyridine ≥97%
6-Chloro-2-fluoro-3-(methylthio)pyridine

Precaución: For research use only

Formula: C₆H₅ClFNS
MW: 177.627 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30469195
CAS Number: 2091530-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR