5-Bromo-2-ethoxy-1-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53366-500MG 53366-5G 53366-25G
77484-424EA 285.31 USD
77484-424 77494-932 77504-414
5-Bromo-2-ethoxy-1-fluoro-3-methoxybenzene ≥97%
5-Bromo-2-ethoxy-1-fluoro-3-methoxybenzene

Precaución: For research use only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404128
CAS Number: 2385830-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR