Ethyl 3-bromo-2-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

53225-500MG 53225-5G 53225-25G
77484-420EA 128.75 USD
77484-420 77494-928 77504-410
Ethyl 3-bromo-2-(trifluoromethoxy)benzoate ≥97%
Ethyl 3-bromo-2-(trifluoromethoxy)benzoate

Precaución: For research use only

Formula: C₁₀H₈BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28746660
CAS Number: 1804402-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR