1,3-Di-tert-butyl-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

53099-500MG 53099-5G 53099-25G
77484-412EA 87.55 USD
77484-412 77494-918 77504-404
1,3-Di-tert-butyl-5-iodobenzene ≥97%
1,3-Di-tert-butyl-5-iodobenzene

Precaución: For research use only

Formula: C₁₄H₂₁I
Storage Temperature: Ambient
MDL Number: MFCD16875483
CAS Number: 37055-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR