2-(3-Bromo-2,6-dichlorophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

53069-500MG 53069-5G 53069-25G
77484-408EA 260.59 USD
77484-408 77494-914 77504-400
2-(3-Bromo-2,6-dichlorophenyl)-1,3-dioxolane ≥95%
2-(3-Bromo-2,6-dichlorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrCl₂O₂
MW: 297.961 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404116
CAS Number: 2635937-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR