2-(4-Bromo-3-methylphenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

52994-500MG 52994-5G 52994-25G
77484-404EA 95.79 USD
77484-404 77494-908 77504-394
2-(4-Bromo-3-methylphenyl)-2-methyl-1,3-dioxolane ≥97%
2-(4-Bromo-3-methylphenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404104
CAS Number: 56256-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR