3,4-Difluoro-2-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52946-500MG 52946-5G 52946-25G
77484-400EA 136.99 USD
77484-400 77494-904 77504-390
3,4-Difluoro-2-(methylthio)benzaldehyde ≥97%
3,4-Difluoro-2-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₈H₆F₂OS
MW: 188.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30615969
CAS Number: 2274030-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR